Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Bromo-1-chloro-4-fluorobenzene, 98+%

Click to view available options
Quantity:
5g
25g
201849-15-2
C6H3BrClF
209.442
MFCD00061120
FOCCSIJMXBTKHD-UHFFFAOYSA-N
2-chloro-5-fluorobromobenzene,1-bromo-2-chloro-5-fluorobenzene,3-bromo-4-chlorofluorobenzene,2-bromo-1-chloro-4-fluoro-benzene,benzene, 2-bromo-1-chloro-4-fluoro,pubchem4270,acmc-209f6i,ksc494o8h,1-chloro-2-bromo-4-fluorobenzene,2-bromo-4-fluorochlorobenzene
2773263
2-bromo-1-chloro-4-fluorobenzene
C1=CC(=C(C=C1F)Br)Cl
201849-15-2
C6H3BrClF
209.442
MFCD00061120
FOCCSIJMXBTKHD-UHFFFAOYSA-N
2-chloro-5-fluorobromobenzene,1-bromo-2-chloro-5-fluorobenzene,3-bromo-4-chlorofluorobenzene,2-bromo-1-chloro-4-fluoro-benzene,benzene, 2-bromo-1-chloro-4-fluoro,pubchem4270,acmc-209f6i,ksc494o8h,1-chloro-2-bromo-4-fluorobenzene,2-bromo-4-fluorochlorobenzene
2773263
2-bromo-1-chloro-4-fluorobenzene
C1=CC(=C(C=C1F)Br)Cl
Quantity 25g
Formula Weight 209.45
Percent Purity ≥98%
Chemical Name or Material 2-Bromo-1-chloro-4-fluorobenzene

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning