Alfa Aesar™ 2-Bromo-1-(ethoxymethyl)imidazole, 97%

Manufacturer: Alfa Aesar™ H5186003

Quantity 1g

 View more versions of this product

Catalog No. AAH5186003

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 850429-54-8
    Molecular Formula C6H9BrN2O
    Formula Weight 205.05
    MDL Number MFCD04038915
    Chemical Name or Material 2-Bromo-1-(ethoxymethyl)imidazole

    Specifications

    Formula Weight 205.05
    Molecular Formula C6H9BrN2O
    MDL Number MFCD04038915
    Quantity 1g
    Health Hazard 2 GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.
    CAS 850429-54-8
    Health Hazard 1 Warning
    Health Hazard 3 GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Chemical Name or Material 2-Bromo-1-(ethoxymethyl)imidazole
    Assay 97%

    Description

    Applications
    2-Bromo-1-(ethoxymethyl)imidazole is used in pharmaceuticals, drug candidates, ligands for transition metal catalysts and other molecular functional materials.

    Solubility
    Very slightly soluble in water (11 g/L at 25°C).

    Notes
    Stable under recommended storage conditions. Store away from oxidizing agents.

    GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit