Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Bromo-1-naphthol, 95%

Click to view available options
Quantity:
250mg
1g
5g
771-15-3
C10H7BrO
223.069
MFCD00274158
LOHOSHDZQVKDPS-UHFFFAOYSA-N
2-bromo-1-naphthol,2-bromo-naphthalen-1-ol,2-bromo-1-naphthalenol,1-naphthalenol, 2-bromo,naphthalenol, bromo,bromonaphthol,bromnaphthol,pubchem22909,2-bromanylnaphthalen-1-ol,acmc-209vk0
150849
2-bromonaphthalen-1-ol
C1=CC=C2C(=C1)C=CC(=C2O)Br
771-15-3
C10H7BrO
223.069
MFCD00274158
LOHOSHDZQVKDPS-UHFFFAOYSA-N
2-bromo-1-naphthol,2-bromo-naphthalen-1-ol,2-bromo-1-naphthalenol,1-naphthalenol, 2-bromo,naphthalenol, bromo,bromonaphthol,bromnaphthol,pubchem22909,2-bromanylnaphthalen-1-ol,acmc-209vk0
150849
2-bromonaphthalen-1-ol
C1=CC=C2C(=C1)C=CC(=C2O)Br
Color White
Quantity 250mg
Beilstein 2554660
Formula Weight 223.07
Physical Form Powder
Percent Purity 95%
Chemical Name or Material 2-Bromo-1-naphthol

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : -30° to -10°C