Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-2',4'-dichloroacetophenone, 97%, ACROS Organics™

Click to view available options
Quantity:
25g
Packaging:
Glass bottle
2631-72-3
C8H5BrCl2O
267.931
MFCD00053005
DASJDMQCPIDJIF-UHFFFAOYSA-N
2-bromo-2',4'-dichloroacetophenone,2-bromo-1-2,4-dichlorophenyl ethanone,2,4-dichlorophenacyl bromide,2-bromo-1-2,4-dichlorophenyl ethan-1-one,2-bromo-1-2,4-dichloro-phenyl-ethanone,ethanone, 2-bromo-1-2,4-dichlorophenyl,2'-bromo-2,4-dichloro acetophenone,omega-bromo-2,4-dichloro acetophenone,2',4'-dichlorophenacyl bromide,2-bromo-2,4-dichloroacetophenone
75828
2-bromo-1-(2,4-dichlorophenyl)ethanone
C1=CC(=C(C=C1Cl)Cl)C(=O)CBr
2631-72-3
C8H5BrCl2O
267.931
MFCD00053005
DASJDMQCPIDJIF-UHFFFAOYSA-N
2-bromo-2',4'-dichloroacetophenone,2-bromo-1-2,4-dichlorophenyl ethanone,2,4-dichlorophenacyl bromide,2-bromo-1-2,4-dichlorophenyl ethan-1-one,2-bromo-1-2,4-dichloro-phenyl-ethanone,ethanone, 2-bromo-1-2,4-dichlorophenyl,2'-bromo-2,4-dichloro acetophenone,omega-bromo-2,4-dichloro acetophenone,2',4'-dichlorophenacyl bromide,2-bromo-2,4-dichloroacetophenone
75828
2-bromo-1-(2,4-dichlorophenyl)ethanone
C1=CC(=C(C=C1Cl)Cl)C(=O)CBr
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 25g
Color White to Brown
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 267.94
Physical Form Low Melting Solid
Percent Purity 97%
Chemical Name or Material 2-Bromo-2′,4′-dichloroacetophenone, 98%
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger