Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Bromo-2',4'-dimethoxyacetophenone, 98%

Click to view available options
Quantity:
1g
5g
25g
60965-26-6
C10H11BrO3
259.099
MFCD00000197
PKVBZABQCCQHLD-UHFFFAOYSA-N
2-bromo-1-2,4-dimethoxyphenyl ethanone,2-bromo-2',4'-dimethoxyacetophenone,2,4-dimethoxyphenacyl bromide,2-bromo-1-2',4'-dimethoxyphenyl ethanone,2-bromo-1-2,4-dimethoxyphenyl ethan-1-one,ethanone, 2-bromo-1-2,4-dimethoxyphenyl,2-bromo-2',4'-dimethoxyacetopheneone,1-2,4-dimethoxyphenyl-2-bromoethan-1-one,pubchem13433,acmc-1ba4l
98683
2-bromo-1-(2,4-dimethoxyphenyl)ethanone
COC1=CC(=C(C=C1)C(=O)CBr)OC
60965-26-6
C10H11BrO3
259.099
MFCD00000197
PKVBZABQCCQHLD-UHFFFAOYSA-N
2-bromo-1-2,4-dimethoxyphenyl ethanone,2-bromo-2',4'-dimethoxyacetophenone,2,4-dimethoxyphenacyl bromide,2-bromo-1-2',4'-dimethoxyphenyl ethanone,2-bromo-1-2,4-dimethoxyphenyl ethan-1-one,ethanone, 2-bromo-1-2,4-dimethoxyphenyl,2-bromo-2',4'-dimethoxyacetopheneone,1-2,4-dimethoxyphenyl-2-bromoethan-1-one,pubchem13433,acmc-1ba4l
98683
2-bromo-1-(2,4-dimethoxyphenyl)ethanone
COC1=CC(=C(C=C1)C(=O)CBr)OC
Melting Point 102°C to 105°C
Quantity 25g
UN Number UN3261
Formula Weight 259.1
Percent Purity 98%
Chemical Name or Material 2-Bromo-2',4'-dimethoxyacetophenone

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

Recommended Storage : 2° to 8°C