Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-2',5'-dimethoxyacetophenone 97%, Acros Organics

Click to view available options
Quantity:
5g
1204-21-3
C10H11BrO3
259.099
MFCD00000198
RGQNFYVSEWDUEI-UHFFFAOYSA-N
2-bromo-1-2,5-dimethoxyphenyl ethanone,2-bromo-2',5'-dimethoxyacetophenone,ethanone, 2-bromo-1-2,5-dimethoxyphenyl,2,5-dimethoxyphenacyl bromide,unii-xv6er0c1dt,bromomethyl 2,5-dimethoxyphenyl ketone,alpha-bromo-2',5'-dimethoxyacetophenone,2-bromo-1-2,5-dimethoxyphenyl ethan-1-one,a-bromo-2',5'-dimethoxyacetophenone,xv6er0c1dt
70997
2-bromo-1-(2,5-dimethoxyphenyl)ethanone
COC1=CC(=C(C=C1)OC)C(=O)CBr
1204-21-3
C10H11BrO3
259.099
MFCD00000198
RGQNFYVSEWDUEI-UHFFFAOYSA-N
2-bromo-1-2,5-dimethoxyphenyl ethanone,2-bromo-2',5'-dimethoxyacetophenone,ethanone, 2-bromo-1-2,5-dimethoxyphenyl,2,5-dimethoxyphenacyl bromide,unii-xv6er0c1dt,bromomethyl 2,5-dimethoxyphenyl ketone,alpha-bromo-2',5'-dimethoxyacetophenone,2-bromo-1-2,5-dimethoxyphenyl ethan-1-one,a-bromo-2',5'-dimethoxyacetophenone,xv6er0c1dt
70997
2-bromo-1-(2,5-dimethoxyphenyl)ethanone
COC1=CC(=C(C=C1)OC)C(=O)CBr
Melting Point 83°C to 88°C
Quantity 5g
Infrared Spectrum Authentic
Color Beige
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula (CH3O)2C6H3COCH2Br
Formula Weight 259.09
Physical Form Fine Crystalline Powder or Needles
Percent Purity ≥96.0%
Chemical Name or Material 2-Bromo-2', 5'-dimethoxyacetophenone, 97%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

LACHRYMATOR

EINECSNumber : 214-873-2

TSCA : TSCA