Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-2'-methoxyacetophenone, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
31949-21-0
C9H9BrO2
229.07
MFCD00000196
GKNCPTLOPRDYMH-UHFFFAOYSA-N
2-bromo-1-2-methoxyphenyl ethanone,2-bromo-2'-methoxyacetophenone,2-methoxyphenacyl bromide,o-methoxy phenacylbromide,bromomethyl 2-methoxyphenyl ketone,2-bromo-1-2-methoxyphenyl ethan-1-one,2-bromo-2'-methoxy acetophenone,ethanone, 2-bromo-1-methoxyphenyl,alpha-bromo-o-methoxyacetophenone,2-bromoacetylanisole
123440
2-bromo-1-(2-methoxyphenyl)ethanone
COC1=CC=CC=C1C(=O)CBr
31949-21-0
C9H9BrO2
229.07
MFCD00000196
GKNCPTLOPRDYMH-UHFFFAOYSA-N
2-bromo-1-2-methoxyphenyl ethanone,2-bromo-2'-methoxyacetophenone,2-methoxyphenacyl bromide,o-methoxy phenacylbromide,bromomethyl 2-methoxyphenyl ketone,2-bromo-1-2-methoxyphenyl ethan-1-one,2-bromo-2'-methoxy acetophenone,ethanone, 2-bromo-1-methoxyphenyl,alpha-bromo-o-methoxyacetophenone,2-bromoacetylanisole
123440
2-bromo-1-(2-methoxyphenyl)ethanone
COC1=CC=CC=C1C(=O)CBr
Color White
Flash Point 113°C
Quantity 1g
Melting Point 43.0°C to 45.0°C
Infrared Spectrum Authentic
Packaging Glass bottle
Boiling Point 130.0°C (1.0 mmHg)
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula CH3OC6H4COCH2Br
Formula Weight 229.07
Physical Form Crystalline Powder or Crystals
Percent Purity 98%
Chemical Name or Material 2-Bromo-2'-methoxyacetophenone
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 250-870-2