Alfa Aesar™ 2-Bromo-3',5'-di-tert-butyl-4'-hydroxyacetophenone, 97%

Manufacturer: Alfa Aesar™ H2683506

Quantity 5g

 View more versions of this product

Catalog No. AAH2683506

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 14386-64-2
    Molecular Formula C16H23BrO2
    Formula Weight 327.26
    MDL Number MFCD00173889
    UN Number UN1759
    Synonym 3,5-Di-tert-butyl-4-hydroxyphenacyl bromide
    Chemical Name or Material 2-Bromo-3',5'-di-tert-butyl-4'-hydroxyacetophenone

    Specifications

    Formula Weight 327.26
    Molecular Formula C16H23BrO2
    MDL Number MFCD00173889
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
    CAS 14386-64-2
    UN Number UN1759
    Melting Point 104° to 107°C
    Synonym 3,5-Di-tert-butyl-4-hydroxyphenacyl bromide
    Quantity 5g
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material 2-Bromo-3',5'-di-tert-butyl-4'-hydroxyacetophenone
    Assay 97%

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit