Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Bromo-3,5-dinitrobenzoic acid, 98%

Click to view available options
Quantity:
1g
5g
25g
116529-60-3
C7H3BrN2O6
291.013
MFCD00458630
BBKJKUFARRJPRP-UHFFFAOYSA-N
2-bromo-3,5-dinitrobenzoic acid,acmc-1bobs,2-bromo-3,5-dinitrobenzoicacid,benzoic acid,2-bromo-3,5-dinitro
3769937
C1=C(C=C(C(=C1[N+](=O)[O-])Br)C(=O)O)[N+](=O)[O-]
116529-60-3
C7H3BrN2O6
291.013
MFCD00458630
BBKJKUFARRJPRP-UHFFFAOYSA-N
2-bromo-3,5-dinitrobenzoic acid,acmc-1bobs,2-bromo-3,5-dinitrobenzoicacid,benzoic acid,2-bromo-3,5-dinitro
3769937
C1=C(C=C(C(=C1[N+](=O)[O-])Br)C(=O)O)[N+](=O)[O-]
Sensitivity Light Sensitive
Quantity 25g
Color White
Melting Point 213°C to 216°C
Beilstein 2146483
Formula Weight 291.01
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 2-Bromo-3,5-dinitrobenzoic acid