Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Bromo-3'-chloroacetophenone, 97%

Click to view available options
Quantity:
1g
5g
25g
41011-01-2
C8H6BrClO
233.489
MFCD00792669
KJVRURZDIOVSSQ-UHFFFAOYSA-N
2-bromo-1-3-chlorophenyl ethanone,3-chlorophenacyl bromide,2-bromo-3'-chloroacetophenone,2-bromo-1-3-chlorophenyl ethan-1-one,2-bromo-1-3-chlorophenyl-1-ethanone,chlorophenacyl bromide,3-chlorophenacylbromide,3'-chlorophenacyl bromide,unii-n62718dtbs,ethanone, 2-bromo-1-3-chlorophenyl
38738
2-bromo-1-(3-chlorophenyl)ethanone
C1=CC(=CC(=C1)Cl)C(=O)CBr
41011-01-2
C8H6BrClO
233.489
MFCD00792669
KJVRURZDIOVSSQ-UHFFFAOYSA-N
2-bromo-1-3-chlorophenyl ethanone,3-chlorophenacyl bromide,2-bromo-3'-chloroacetophenone,2-bromo-1-3-chlorophenyl ethan-1-one,2-bromo-1-3-chlorophenyl-1-ethanone,chlorophenacyl bromide,3-chlorophenacylbromide,3'-chlorophenacyl bromide,unii-n62718dtbs,ethanone, 2-bromo-1-3-chlorophenyl
38738
2-bromo-1-(3-chlorophenyl)ethanone
C1=CC(=CC(=C1)Cl)C(=O)CBr
Applications
2-Bromo-3′-chloroacetophenone acts as a Substrate used in a one-step preparation of symmetrical 1,4-diketones in the presence of Zn-I2.

Notes
Store at 4°C. Incompatible with oxidizing agents.
Boiling Point 122°C to 124°C
Quantity 25g
Melting Point 39°C to 43°C
UN Number UN3335
Beilstein 2086002
Formula Weight 233.49
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 2-Bromo-3'-chloroacetophenone

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 9; Packaging Group: ; Proper Shipping Name: AVIATION REGULATED SOLID, N.O.S.

storageNote1 : 2° to 8°C