Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Bromo-3-chlorobenzotrifluoride, 98%

Click to view available options
Quantity:
250mg
1g
5g
384-16-7
C7H3BrClF3
259.45
MFCD00673988
GMUWNTUONIQRFP-UHFFFAOYSA-N
2-bromo-1-chloro-3-trifluoromethyl benzene,2-bromo-3-chlorobenzotrifluoride,2-bromo-3-chloro-1-trifluoromethyl benzene,2-bromanyl-1-chloranyl-3-trifluoromethyl benzene,benzene, 2-bromo-1-chloro-3-trifluoromethyl
17750574
2-bromo-1-chloro-3-(trifluoromethyl)benzene
C1=CC(=C(C(=C1)Cl)Br)C(F)(F)F
384-16-7
C7H3BrClF3
259.45
MFCD00673988
GMUWNTUONIQRFP-UHFFFAOYSA-N
2-bromo-1-chloro-3-trifluoromethyl benzene,2-bromo-3-chlorobenzotrifluoride,2-bromo-3-chloro-1-trifluoromethyl benzene,2-bromanyl-1-chloranyl-3-trifluoromethyl benzene,benzene, 2-bromo-1-chloro-3-trifluoromethyl
17750574
2-bromo-1-chloro-3-(trifluoromethyl)benzene
C1=CC(=C(C(=C1)Cl)Br)C(F)(F)F
Quantity 5g
Beilstein 21501675
Formula Weight 259.46
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 2-Bromo-3-chlorobenzotrifluoride

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning