Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Bromo-3'-chloropropiophenone, 98%

Click to view available options
Quantity:
1g
5g
34911-51-8
C9H8BrClO
247.516
MFCD03701150
OFNMQTRHMBQQEA-UHFFFAOYSA-N
2-bromo-3'-chloropropiophenone,2-bromo-1-3-chlorophenyl propan-1-one,2-bromo-1-3-chlorophenyl-1-propanone,3'-chloro-2-bromopropiophenone,1-propanone,2-bromo-1-3-chlorophenyl,1-propanone, 2-bromo-1-3-chlorophenyl,2-bromo-1-3-chloro-phenyl-propan-1-one,bromochlorophenylpropanone,bromo-3'-chloropropiophenone,ksc573e4l
4992116
2-bromo-1-(3-chlorophenyl)propan-1-one
CC(C(=O)C1=CC(=CC=C1)Cl)Br
34911-51-8
C9H8BrClO
247.516
MFCD03701150
OFNMQTRHMBQQEA-UHFFFAOYSA-N
2-bromo-3'-chloropropiophenone,2-bromo-1-3-chlorophenyl propan-1-one,2-bromo-1-3-chlorophenyl-1-propanone,3'-chloro-2-bromopropiophenone,1-propanone,2-bromo-1-3-chlorophenyl,1-propanone, 2-bromo-1-3-chlorophenyl,2-bromo-1-3-chloro-phenyl-propan-1-one,bromochlorophenylpropanone,bromo-3'-chloropropiophenone,ksc573e4l
4992116
2-bromo-1-(3-chlorophenyl)propan-1-one
CC(C(=O)C1=CC(=CC=C1)Cl)Br
Applications
It used as an important intermediate for raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in acetonitrile, chloroform, dichloromethane and ethyl acetate.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
Quantity 5g
Color Colorless to Yellow
Density 1.532g/mL
Refractive Index 1.5770
Beilstein 2360404
Formula Weight 247.52
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 2-Bromo-3'-chloropropiophenone

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : 2° to 8°C