Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-3-chloropyridine, 97%, Acros Organics

Click to view available options
Quantity:
1g
5g
96424-68-9
C5H3BrClN
192.44
GOHBBINNYAWQGO-UHFFFAOYSA-N
pyridine, 2-bromo-3-chloro,3-chloro-2-bromopyridine,2-bromo-3-chloro-pyridine,chloro bromo pyridine,pubchem5776,2-bromo-3-pyridyl chloride,ksc486m1b,2-bromo-3-chloropyridine,abbypharma ap-18-5389
7016326
2-bromo-3-chloropyridine
C1=CC(=C(N=C1)Br)Cl
96424-68-9
C5H3BrClN
192.44
GOHBBINNYAWQGO-UHFFFAOYSA-N
pyridine, 2-bromo-3-chloro,3-chloro-2-bromopyridine,2-bromo-3-chloro-pyridine,chloro bromo pyridine,pubchem5776,2-bromo-3-pyridyl chloride,ksc486m1b,2-bromo-3-chloropyridine,abbypharma ap-18-5389
7016326
2-bromo-3-chloropyridine
C1=CC(=C(N=C1)Br)Cl
Melting Point 60.0°C to 62.0°C
Quantity 5g
Assay Percent Range 96% min. (GC)
Formula Weight 192.44
Percent Purity 97%
Chemical Name or Material 2-Bromo-3-chloropyridine

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and kee

GHS Signal Word: Warning