Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Bromo-3-fluoro-5-(trifluoromethyl)pyridine, 97%

Click to view available options
Quantity:
250mg
1g
89402-29-9
C6H2BrF4N
243.987
MFCD10699118
DNVIECHQUYQOOH-UHFFFAOYSA-N
2-bromo-3-fluoro-5-trifluoromethyl pyridine,pyridine,2-bromo-3-fluoro-5-trifluoromethyl,pyridine, 2-bromo-3-fluoro-5-trifluoromethyl,acmc-20a5we,2-bromo-3-fluoro-5-trifluoromethyl-pyridine
15592823
2-bromo-3-fluoro-5-(trifluoromethyl)pyridine
C1=C(C=NC(=C1F)Br)C(F)(F)F
89402-29-9
C6H2BrF4N
243.987
MFCD10699118
DNVIECHQUYQOOH-UHFFFAOYSA-N
2-bromo-3-fluoro-5-trifluoromethyl pyridine,pyridine,2-bromo-3-fluoro-5-trifluoromethyl,pyridine, 2-bromo-3-fluoro-5-trifluoromethyl,acmc-20a5we,2-bromo-3-fluoro-5-trifluoromethyl-pyridine
15592823
2-bromo-3-fluoro-5-(trifluoromethyl)pyridine
C1=C(C=NC(=C1F)Br)C(F)(F)F
Applications
2-Bromo-3-fluoro-5-(trifluoromethyl)pyridine is used as a pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Refractive Index 1.464
Quantity 250mg
Solubility Information Not miscible or difficult to mix.
Formula Weight 243.99
Percent Purity 97%
Chemical Name or Material 2-Bromo-3-fluoro-5-(trifluoromethyl)pyridine

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures