Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Bromo-3-fluoro-6-methylpyridine, 98%

Click to view available options
Quantity:
1g
5g
25g
374633-36-0
C6H5BrFN
190.015
MFCD03095061
KKVUMKHUMBXLMH-UHFFFAOYSA-N
2-bromo-3-fluoro-6-picoline,2-bromo-3-fluoro-6-methyl-pyridine,pubchem3001
24820318
2-bromo-3-fluoro-6-methylpyridine
CC1=NC(=C(C=C1)F)Br
374633-36-0
C6H5BrFN
190.015
MFCD03095061
KKVUMKHUMBXLMH-UHFFFAOYSA-N
2-bromo-3-fluoro-6-picoline,2-bromo-3-fluoro-6-methyl-pyridine,pubchem3001
24820318
2-bromo-3-fluoro-6-methylpyridine
CC1=NC(=C(C=C1)F)Br
Quantity 25g
Beilstein 18506793
Formula Weight 190.02
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 2-Bromo-3-fluoro-6-methylpyridine