Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-3-fluorobenzoic acid, 98%, Acros Organics™

Click to view available options
Quantity:
1g
5g
132715-69-6
C7H4BrFO2
219.01
KQRCBMPPEPNNDS-UHFFFAOYSA-N
3-fluoro-2-bromobenzoic acid,2-bromo-3-fluorobenzoicacid,2-bromo-3-fluoro-benzoic acid,buttpark 22\01-97,benzoic acid, 2-bromo-3-fluoro,pubchem1319,2-bromo-3-fluorobenzoic,acmc-1c0re,intermediates-zcf02016,ksc174c5p
302622
2-bromo-3-fluorobenzoic acid
C1=CC(=C(C(=C1)F)Br)C(=O)O
132715-69-6
C7H4BrFO2
219.01
KQRCBMPPEPNNDS-UHFFFAOYSA-N
3-fluoro-2-bromobenzoic acid,2-bromo-3-fluorobenzoicacid,2-bromo-3-fluoro-benzoic acid,buttpark 22\01-97,benzoic acid, 2-bromo-3-fluoro,pubchem1319,2-bromo-3-fluorobenzoic,acmc-1c0re,intermediates-zcf02016,ksc174c5p
302622
2-bromo-3-fluorobenzoic acid
C1=CC(=C(C(=C1)F)Br)C(=O)O
Infrared Spectrum Authentic
Melting Point 162.0°C to 164.0°C
Quantity 5g
Assay Percent Range 97.5% min. (HPLC)
Formula Weight 219.01
Percent Purity 98%
Chemical Name or Material 2-Bromo-3-fluorobenzoic acid

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning