Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Bromo-3-fluorobenzotrifluoride, 97%

Click to view available options
Quantity:
1g
5g
104540-42-3
C7H3BrF4
242.999
MFCD00061243
UERAGXKMOXUWPC-UHFFFAOYSA-N
2-bromo-3-fluorobenzotrifluoride,2-bromo-1-fluoro-3-trifluoromethyl benzene,2-fluoro-6-trifluoromethyl bromobenzene,2-bromo-3-fluoro-1-trifluoromethyl benzene,benzene, 2-bromo-1-fluoro-3-trifluoromethyl,2-bromo-1-fluoro-3-trifluoromethyl-benzene,2-bromo-3-fluorobenzo trifluoride,2-bromo-3-fluorbenzotrifluoride,2-fluoro-6-trifluoromethyl-bromobenzene
517847
2-bromo-1-fluoro-3-(trifluoromethyl)benzene
C1=CC(=C(C(=C1)F)Br)C(F)(F)F
104540-42-3
C7H3BrF4
242.999
MFCD00061243
UERAGXKMOXUWPC-UHFFFAOYSA-N
2-bromo-3-fluorobenzotrifluoride,2-bromo-1-fluoro-3-trifluoromethyl benzene,2-fluoro-6-trifluoromethyl bromobenzene,2-bromo-3-fluoro-1-trifluoromethyl benzene,benzene, 2-bromo-1-fluoro-3-trifluoromethyl,2-bromo-1-fluoro-3-trifluoromethyl-benzene,2-bromo-3-fluorobenzo trifluoride,2-bromo-3-fluorbenzotrifluoride,2-fluoro-6-trifluoromethyl-bromobenzene
517847
2-bromo-1-fluoro-3-(trifluoromethyl)benzene
C1=CC(=C(C(=C1)F)Br)C(F)(F)F
Quantity 1g
Formula Weight 243
Percent Purity 97%
Chemical Name or Material 2-Bromo-3-fluorobenzotrifluoride

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning