Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Bromo-3-fluoropyridine-4-carboxylic acid, 98%

Click to view available options
Quantity:
250mg
1g
5g
1211530-89-0
C6H3BrFNO2
219.997
MFCD13185796
DHFIVEUMGCJILQ-UHFFFAOYSA-N
2-bromo-3-fluoroisonicotinic acid,2-bromo-3-fluoroisonicotinicacid,2-bromo-3-fluoro-4-picolic acid,2-bromo-3-fluoro-4-pyridinecarboxylic acid,2-bromo-3-fluoropyridin-4-carboxylicacid,2-bromo-3-fluoropyridin-4-carboxylic acid
70680697
2-bromo-3-fluoropyridine-4-carboxylic acid
C1=CN=C(C(=C1C(=O)O)F)Br
1211530-89-0
C6H3BrFNO2
219.997
MFCD13185796
DHFIVEUMGCJILQ-UHFFFAOYSA-N
2-bromo-3-fluoroisonicotinic acid,2-bromo-3-fluoroisonicotinicacid,2-bromo-3-fluoro-4-picolic acid,2-bromo-3-fluoro-4-pyridinecarboxylic acid,2-bromo-3-fluoropyridin-4-carboxylicacid,2-bromo-3-fluoropyridin-4-carboxylic acid
70680697
2-bromo-3-fluoropyridine-4-carboxylic acid
C1=CN=C(C(=C1C(=O)O)F)Br
Quantity 250mg
Formula Weight 220
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 2-Bromo-3-fluoropyridine-4-carboxylic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning