Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-3'-methoxyacetophenone 97%, ACROS Organics™

Click to view available options
Quantity:
10g
25g
Packaging:
Glass bottle
5000-65-7
C9H9BrO2
229.07
MFCD00000199
IOOHBIFQNQQUFI-UHFFFAOYSA-N
2-bromo-1-3-methoxyphenyl ethanone,2-bromo-3'-methoxyacetophenone,m-methoxyphenacyl bromide,3'-methoxyphenacyl bromide,3-bromoacetyl anisole,2-bromo-1-3-methoxyphenyl ethan-1-one,3-methoxyphenacyl bromide,bromomethyl 3-methoxyphenyl ketone,ccris 7998,3-methoxyphenacylbromide
101294
2-bromo-1-(3-methoxyphenyl)ethanone
COC1=CC=CC(=C1)C(=O)CBr
5000-65-7
C9H9BrO2
229.07
MFCD00000199
IOOHBIFQNQQUFI-UHFFFAOYSA-N
2-bromo-1-3-methoxyphenyl ethanone,2-bromo-3'-methoxyacetophenone,m-methoxyphenacyl bromide,3'-methoxyphenacyl bromide,3-bromoacetyl anisole,2-bromo-1-3-methoxyphenyl ethan-1-one,3-methoxyphenacyl bromide,bromomethyl 3-methoxyphenyl ketone,ccris 7998,3-methoxyphenacylbromide
101294
2-bromo-1-(3-methoxyphenyl)ethanone
COC1=CC=CC(=C1)C(=O)CBr
Melting Point 59°C to 64°C
Packaging Glass bottle
Color Green-Yellow to White
Quantity 25g
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula CH3OC6H4COCH2Br
Formula Weight 229.07
Physical Form Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material 2-Bromo-3'-methoxyacetophenone, 97%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 225-666-1

TSCA : TSCA