Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-3-methoxypyridine, 97%, ACROS Organics™

Click to view available options
Quantity:
10g
24100-18-3
C6H6BrNO
188.02
MFCD01570896
PDOWLYNSFYZIQX-UHFFFAOYSA-N
2-bromo-3-methoxy-pyridine,pyridine, 2-bromo-3-methoxy,2-bromo-3-methoxy pyridine,pubchem5916,acmc-1cefu,ksc552s2b,#,pyridine,2-bromo-3-methoxy,2-bromo-3-methoxypyridine,abbypharma ap-17-5948
90364
2-bromo-3-methoxypyridine
COC1=C(N=CC=C1)Br
24100-18-3
C6H6BrNO
188.02
MFCD01570896
PDOWLYNSFYZIQX-UHFFFAOYSA-N
2-bromo-3-methoxy-pyridine,pyridine, 2-bromo-3-methoxy,2-bromo-3-methoxy pyridine,pubchem5916,acmc-1cefu,ksc552s2b,#,pyridine,2-bromo-3-methoxy,2-bromo-3-methoxypyridine,abbypharma ap-17-5948
90364
2-bromo-3-methoxypyridine
COC1=C(N=CC=C1)Br
Packaging Glass bottle
Infrared Spectrum Authentic
Melting Point 45.0°C to 47.0°C
Quantity 10g
Color White to Cream
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Formula Weight 188.02
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material 2-Bromo-3-methoxypyridine
Show More Show Less

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye damage.
Harmful if inhaled.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Danger

EINECSNumber : 246-017-9