Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Bromo-3-methylbutyraldehyde, 96%

Manufacturer:  Alfa Aesar™ H5233406

Quantity 5g

 View more versions of this product

Catalog No. AAH5233406

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Description

Applications
2-Bromo-3-methylbutyraldehyde is used for proteomic research.

Solubility
Miscible with water.

Notes
Incompatible with strong oxidizing agents and strong acids. Air sensitive.
Description & Specifications

Chemical Identifiers

C5H9BrO
165.03
UN1989
(CH3)2CHCH(Br)CHO
MFCD11846369
2-Bromo-3-methylbutyraldehyde

Specifications

C5H9BrO
165.03
UN1989
45°C (113°F)
96%
(CH3)2CHCH(Br)CHO
MFCD11846369
5g
2-Bromo-3-methylbutyraldehyde

Safety and Handling

SDS & Safety Documents

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALDEHYDES, N.O.S.

Recommended Storage : 2° to 8°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit