Alfa Aesar™ 2-Bromo-3-methylbutyraldehyde, 96%

Manufacturer: Alfa Aesar™ H52334MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH52334MD

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    Molecular Formula C5H9BrO
    Linear Formula (CH3)2CHCH(Br)CHO
    Formula Weight 165.03
    MDL Number MFCD11846369
    UN Number UN1989
    Chemical Name or Material 2-Bromo-3-methylbutyraldehyde

    Specifications

    Formula Weight 165.03
    Molecular Formula C5H9BrO
    Linear Formula (CH3)2CHCH(Br)CHO
    MDL Number MFCD11846369
    UN Number UN1989
    Quantity 250mg
    Flash Point 45°C (113°F)
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Recommended Storage 2° to 8°C
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALDEHYDES, N.O.S.
    Chemical Name or Material 2-Bromo-3-methylbutyraldehyde
    Assay 96%

    Description

    Applications
    2-Bromo-3-methylbutyraldehyde is used for proteomic research.

    Solubility
    Miscible with water.

    Notes
    Incompatible with strong oxidizing agents and strong acids. Air sensitive.

    GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    2° to 8°C

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit