Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-3-pyridinol 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
6602-32-0
C5H4BrNO
174
MFCD00006220
YKHQFTANTNMYPP-UHFFFAOYSA-N
2-bromo-3-hydroxypyridine,2-bromo-3-pyridinol,3-pyridinol, 2-bromo,2-bromo-pyridin-3-ol,3-hydroxy-2-bromopyridine,unii-9dab85h359,2-bromo-3-hydroxy pyridine,2-bromo-3-hydroxy-pyridine,2-bromo-3-hydroxypyridine 2-bromo-3-pyridinol,2-bromo3-pyridinol
23070
2-bromopyridin-3-ol
C1=CC(=C(N=C1)Br)O
6602-32-0
C5H4BrNO
174
MFCD00006220
YKHQFTANTNMYPP-UHFFFAOYSA-N
2-bromo-3-hydroxypyridine,2-bromo-3-pyridinol,3-pyridinol, 2-bromo,2-bromo-pyridin-3-ol,3-hydroxy-2-bromopyridine,unii-9dab85h359,2-bromo-3-hydroxy pyridine,2-bromo-3-hydroxy-pyridine,2-bromo-3-hydroxypyridine 2-bromo-3-pyridinol,2-bromo3-pyridinol
23070
2-bromopyridin-3-ol
C1=CC(=C(N=C1)Br)O
Melting Point 185°C to 188°C
Infrared Spectrum Authentic
Quantity 25g
Packaging Glass bottle
Color Brown to Gray
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula Br(C5H3N)OH
Formula Weight 174
Physical Form Crystalline Powder
Percent Purity ≥98.5%
Chemical Name or Material 2-Bromo-3-pyridinol, 99%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 229-547-5

RTECSNumber : UU7705000

TSCA : TSCA