Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-4,5-difluoroanisole, 98%, Alfa Aesar™

Click to view available options
Quantity:
2g
10g
202865-58-5
C7H5BrF2O
223.017
MFCD00070753
DKARISQFDBAJEZ-UHFFFAOYSA-N
2-bromo-4,5-difluoroanisole,benzene, 1-bromo-4,5-difluoro-2-methoxy,4-bromo-1,2-difluoro-5-methoxybenzene,pubchem5223,acmc-1co9o,2-bromo4,5-difluoro-anisole,ksc495e7d,2-bromo-4,5-difluoro-anisole,1-bromo-4,5-difluoro-2-methoxy-benzene,1-bromanyl-4,5-bis fluoranyl-2-methoxy-benzene
2736262
1-bromo-4,5-difluoro-2-methoxybenzene
COC1=CC(=C(C=C1Br)F)F
202865-58-5
C7H5BrF2O
223.017
MFCD00070753
DKARISQFDBAJEZ-UHFFFAOYSA-N
2-bromo-4,5-difluoroanisole,benzene, 1-bromo-4,5-difluoro-2-methoxy,4-bromo-1,2-difluoro-5-methoxybenzene,pubchem5223,acmc-1co9o,2-bromo4,5-difluoro-anisole,ksc495e7d,2-bromo-4,5-difluoro-anisole,1-bromo-4,5-difluoro-2-methoxy-benzene,1-bromanyl-4,5-bis fluoranyl-2-methoxy-benzene
2736262
1-bromo-4,5-difluoro-2-methoxybenzene
COC1=CC(=C(C=C1Br)F)F
Applications
It is employed as a intermediate for pharmaceuticals.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Quantity 2g
Formula Weight 223.02
Percent Purity 98%
Chemical Name or Material 2-Bromo-4,5-difluoroanisole

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning