Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Bromo-4,6-difluoroaniline, 98%

Manufacturer:  Alfa Aesar™ A1102236

Quantity 500g

 View more versions of this product

Catalog No. AAA1102236


  Check Availability
Add to cart

Specifications

Description

Applications
2-Bromo-4,6-difluoroaniline is used in chemical synthesis.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Description & Specifications

Chemical Identifiers

444-14-4
208.01
UN3077
2-Bromo-4,6-difluoroaniline
C6H4BrF2N
MFCD00009639
2718139

Specifications

444-14-4
208.01
UN3077
2718139
Weak, characteristic
98%
C6H4BrF2N
MFCD00009639
38° to 42°C
500g
2-Bromo-4,6-difluoroaniline

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301
Toxic if swallowed.

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit