Alfa Aesar™ 2-Bromo-4,6-difluoroanisole, 97%

Manufacturer: Alfa Aesar™ H2619906

Quantity 5g

 View more versions of this product

Catalog No. AAH2619906

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 202865-59-6
    Molecular Formula C7H5BrF2O
    Formula Weight 223.02
    MDL Number MFCD00070754
    Chemical Name or Material 2-Bromo-4,6-difluoroanisole

    Specifications

    Formula Weight 223.02
    Molecular Formula C7H5BrF2O
    MDL Number MFCD00070754
    Quantity 5g
    Health Hazard 2 GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.
    CAS 202865-59-6
    Health Hazard 1 Warning
    Health Hazard 3 GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Chemical Name or Material 2-Bromo-4,6-difluoroanisole
    Assay 97%

    GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit