Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Bromo-4,6-difluorophenol, 98+%

Click to view available options
Quantity:
5g
25g
98130-56-4
C6H3BrF2O
208.99
MFCD00070752
YDRZYJATKMBSJX-UHFFFAOYSA-N
2-bromo-4,6-difluoro-phenol,buttpark 20\04-78,phenol, 2-bromo-4,6-difluoro,pubchem2836,acmc-209s9n,2-bromo-4,6-difluorophenol,2-bromanyl-4,6-bis fluoranyl phenol,2-bromo-4,6-difluorophenol, 98+%
2736290
2-bromo-4,6-difluorophenol
C1=C(C=C(C(=C1F)O)Br)F
98130-56-4
C6H3BrF2O
208.99
MFCD00070752
YDRZYJATKMBSJX-UHFFFAOYSA-N
2-bromo-4,6-difluoro-phenol,buttpark 20\04-78,phenol, 2-bromo-4,6-difluoro,pubchem2836,acmc-209s9n,2-bromo-4,6-difluorophenol,2-bromanyl-4,6-bis fluoranyl phenol,2-bromo-4,6-difluorophenol, 98+%
2736290
2-bromo-4,6-difluorophenol
C1=C(C=C(C(=C1F)O)Br)F
Flash Point 105°C (221°F)
Boiling Point 53°C (1mmHg)
Quantity 5g
Beilstein 3245985
Formula Weight 208.99
Percent Purity ≥98%
Chemical Name or Material 2-Bromo-4,6-difluorophenol

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning