Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-4-chloro-1-iodobenzene, 98%, Acros Organics

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
31928-44-6
C6H3BrClI
317.36
CXHXFDQEFKFYQJ-UHFFFAOYSA-N
2-bromo-4-chloroiodobenzene,benzene, 2-bromo-4-chloro-1-iodo,acmc-209hpo,ksc494q6f,1-iodo-2-bromo-4-chlorobenzene,2-bromo-1-iodo-4-chlorobenzene
13741326
2-bromo-4-chloro-1-iodobenzene
C1=CC(=C(C=C1Cl)Br)I
31928-44-6
C6H3BrClI
317.36
CXHXFDQEFKFYQJ-UHFFFAOYSA-N
2-bromo-4-chloroiodobenzene,benzene, 2-bromo-4-chloro-1-iodo,acmc-209hpo,ksc494q6f,1-iodo-2-bromo-4-chlorobenzene,2-bromo-1-iodo-4-chlorobenzene
13741326
2-bromo-4-chloro-1-iodobenzene
C1=CC(=C(C=C1Cl)Br)I
Quantity 5g
Packaging Glass bottle
Assay Percent Range 97.5% min. (GC)
Linear Formula BrC6H3(Cl)I
Solubility Information Solubility in water: insoluble
Formula Weight 317.36
Percent Purity 98%
Chemical Name or Material 2-Bromo-4-chloro-1-iodobenzene

GHS Signal Word: Warning