Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-4'-chloroacetophenone, 98+%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
536-38-9
C8H6BrClO
233.49
MFCD00000625
FLAYZKKEOIAALB-UHFFFAOYSA-N
2-bromo-4'-chloroacetophenone,2-bromo-1-4-chlorophenyl ethanone,4-chlorophenacyl bromide,p-chlorophenacyl bromide,2-bromo-1-4-chlorophenyl ethan-1-one,ethanone, 2-bromo-1-4-chlorophenyl,4'-chlorophenacyl bromide,2-bromo-p-chloroacetophenone,p-bromoacetyl chlorobenzene,4'-chloro-2-bromoacetophenone
68303
2-bromo-1-(4-chlorophenyl)ethanone
C1=CC(=CC=C1C(=O)CBr)Cl
536-38-9
C8H6BrClO
233.49
MFCD00000625
FLAYZKKEOIAALB-UHFFFAOYSA-N
2-bromo-4'-chloroacetophenone,2-bromo-1-4-chlorophenyl ethanone,4-chlorophenacyl bromide,p-chlorophenacyl bromide,2-bromo-1-4-chlorophenyl ethan-1-one,ethanone, 2-bromo-1-4-chlorophenyl,4'-chlorophenacyl bromide,2-bromo-p-chloroacetophenone,p-bromoacetyl chlorobenzene,4'-chloro-2-bromoacetophenone
68303
2-bromo-1-(4-chlorophenyl)ethanone
C1=CC(=CC=C1C(=O)CBr)Cl
Infrared Spectrum Authentic
Loss on Drying 1% max. (70°C, 2 hr.)
Color White to Yellow
Melting Point 95.0°C to 99.0°C
Packaging Glass bottle
Quantity 100g
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98+%
Linear Formula ClC6H4COCH2Br
Beilstein 07, 285
Merck Index 15, 2156
Formula Weight 233.49
Physical Form Crystals
Percent Purity ≥98%
Chemical Name or Material 2-Bromo-4′-chloroacetophenone, 98+%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 208-631-5

RTECSNumber : AM5978800

TSCA : TSCA