Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Bromo-4-chlorotoluene, 98%

Click to view available options
Quantity:
5g
25g
100g
27139-97-5
C7H6BrCl
205.479
MFCD00060650
CSUUXPHPCXHYGY-UHFFFAOYSA-N
2-bromo-4-chlorotoluene,benzene, 2-bromo-4-chloro-1-methyl,4-chloro-2-bromotoluene,toluene, 2-bromo-4-chloro,p-chlor brom toluol,pubchem3599,acmc-209gvg,2-bromo-4-chloro toluene,2-bromo-4-chloro-toluene,ksc493k9b
609898
2-bromo-4-chloro-1-methylbenzene
CC1=C(C=C(C=C1)Cl)Br
27139-97-5
C7H6BrCl
205.479
MFCD00060650
CSUUXPHPCXHYGY-UHFFFAOYSA-N
2-bromo-4-chlorotoluene,benzene, 2-bromo-4-chloro-1-methyl,4-chloro-2-bromotoluene,toluene, 2-bromo-4-chloro,p-chlor brom toluol,pubchem3599,acmc-209gvg,2-bromo-4-chloro toluene,2-bromo-4-chloro-toluene,ksc493k9b
609898
2-bromo-4-chloro-1-methylbenzene
CC1=C(C=C(C=C1)Cl)Br
Quantity 25g
Formula Weight 205.48
Percent Purity 98%
Chemical Name or Material 2-Bromo-4-chlorotoluene

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

TSCA : No

Recommended Storage : Ambient temperatures