Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-4'-cyanoacetophenone, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
20099-89-2
C9H6BrNO
224.06
MFCD00052931
LJANCPRIUMHGJE-UHFFFAOYSA-N
4-2-bromoacetyl benzonitrile,4-cyanophenacyl bromide,4-bromoacetyl benzonitrile,2-bromo-4'-cyanoacetophenone,benzonitrile, 4-bromoacetyl,p-cyanophenacyl bromide,4-2-bromo-acetyl-benzonitrile,4-2-bromoethanoyl benzonitrile,4-2-bromoacetyl benzenecarbonitrile
98654
4-(2-bromoacetyl)benzonitrile
C1=CC(=CC=C1C#N)C(=O)CBr
20099-89-2
C9H6BrNO
224.06
MFCD00052931
LJANCPRIUMHGJE-UHFFFAOYSA-N
4-2-bromoacetyl benzonitrile,4-cyanophenacyl bromide,4-bromoacetyl benzonitrile,2-bromo-4'-cyanoacetophenone,benzonitrile, 4-bromoacetyl,p-cyanophenacyl bromide,4-2-bromo-acetyl-benzonitrile,4-2-bromoethanoyl benzonitrile,4-2-bromoacetyl benzenecarbonitrile
98654
4-(2-bromoacetyl)benzonitrile
C1=CC(=CC=C1C#N)C(=O)CBr
Color White to Yellow
Packaging Glass bottle
Quantity 5g
Melting Point 85.0°C to 88.0°C
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Linear Formula BrCH2COC6H4CN
Formula Weight 224.06
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material 2-Bromo-4'-cyanoacetophenone, 97%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger