Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2'-Bromo-4'-fluoroacetanilide, 98%

Click to view available options
Quantity:
5g
25g
100g
1009-22-9
C8H7BrFNO
232.052
MFCD00061118
JAVSBNOXENOHEI-UHFFFAOYSA-N
2'-bromo-4'-fluoroacetanilide,n-2-bromo-4-fluorophenyl acetamide,2-bromo-4-fluoroacetanilide,n-acetyl 2-bromo-4-fluoroaniline,n-2-bromo-4-fluoro-phenyl acetamide,acetamide, n-2-bromo-4-fluorophenyl,maybridge3_000074,ksc180o6b,javsbnoxenohei-uhfffaoysa,n-acetyl2-bromo-4-fluoroaniline
688184
N-(2-bromo-4-fluorophenyl)acetamide
CC(=O)NC1=C(C=C(C=C1)F)Br
1009-22-9
C8H7BrFNO
232.052
MFCD00061118
JAVSBNOXENOHEI-UHFFFAOYSA-N
2'-bromo-4'-fluoroacetanilide,n-2-bromo-4-fluorophenyl acetamide,2-bromo-4-fluoroacetanilide,n-acetyl 2-bromo-4-fluoroaniline,n-2-bromo-4-fluoro-phenyl acetamide,acetamide, n-2-bromo-4-fluorophenyl,maybridge3_000074,ksc180o6b,javsbnoxenohei-uhfffaoysa,n-acetyl2-bromo-4-fluoroaniline
688184
N-(2-bromo-4-fluorophenyl)acetamide
CC(=O)NC1=C(C=C(C=C1)F)Br
Quantity 100g
Melting Point 116°C to 119°C
Beilstein 2803940
Formula Weight 232.05
Percent Purity 98%
Chemical Name or Material 2'-Bromo-4'-fluoroacetanilide

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger