Alfa Aesar™ 2-Bromo-4-fluoroaniline, 98%

Manufacturer: Alfa Aesar™ A1175122

Quantity 100g

 View more versions of this product

Catalog No. AAA1175122

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1003-98-1
    Molecular Formula C6H5BrFN
    Formula Weight 190.01
    MDL Number MFCD00042462
    UN Number UN2810
    Beilstein 2802562
    Chemical Name or Material 2-Bromo-4-fluoroaniline

    Specifications

    Formula Weight 190.01
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Boiling Point 221°C
    Quantity 100g
    Molecular Formula C6H5BrFN
    CAS 1003-98-1
    UN Number UN2810
    Beilstein 2802562
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
    Flash Point 104°C (219°F)
    MDL Number MFCD00042462
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H331-H302-H312-H315-H319
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    Chemical Name or Material 2-Bromo-4-fluoroaniline
    Assay 98%

    GHS H Statement
    H331-H302-H312-H315-H319
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit