Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Bromo-4-fluoropyridine, 97%

Click to view available options
Quantity:
1g
5g
357927-50-5
C5H3BrFN
175.988
MFCD09260909
AIEATTRZKVGMBO-UHFFFAOYSA-N
2-bromo-4-fluoro-pyridine,pyridine, 2-bromo-4-fluoro,4-fluoro-2-bromopyridine,pubchem3004,acmc-209ij7,pybr71,ksc493m3n,2-bromo-4-fluoropyridine,abbypharma ap-17-5103
21885884
2-bromo-4-fluoropyridine
C1=CN=C(C=C1F)Br
357927-50-5
C5H3BrFN
175.988
MFCD09260909
AIEATTRZKVGMBO-UHFFFAOYSA-N
2-bromo-4-fluoro-pyridine,pyridine, 2-bromo-4-fluoro,4-fluoro-2-bromopyridine,pubchem3004,acmc-209ij7,pybr71,ksc493m3n,2-bromo-4-fluoropyridine,abbypharma ap-17-5103
21885884
2-bromo-4-fluoropyridine
C1=CN=C(C=C1F)Br
Quantity 5g
Refractive Index 1.5370
Density 1.699g/mL
Beilstein 14158224
Formula Weight 175.99
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 2-Bromo-4-fluoropyridine

GHS H Statement
H302-H312-H332-H315-H319-H335-H227
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : 2° to 8°C