Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-4'-methoxyacetophenone, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
2632-13-5
C9H9BrO2
229.07
MFCD00000201
XQJAHBHCLXUGEP-UHFFFAOYSA-N
2-bromo-4'-methoxyacetophenone,2-bromo-1-4-methoxyphenyl ethanone,ethanone, 2-bromo-1-4-methoxyphenyl,p-methoxyphenacyl bromide,4'-methoxyphenacyl bromide,2-bromo-1-4-methoxyphenyl ethan-1-one,4-methoxyphenacyl bromide,bromomethyl 4-methoxyphenyl ketone,alpha-bromo-4-methoxyacetophenone,2-bromo-4-methoxyacetophenone
4965
2-bromo-1-(4-methoxyphenyl)ethanone
COC1=CC=C(C=C1)C(=O)CBr
2632-13-5
C9H9BrO2
229.07
MFCD00000201
XQJAHBHCLXUGEP-UHFFFAOYSA-N
2-bromo-4'-methoxyacetophenone,2-bromo-1-4-methoxyphenyl ethanone,ethanone, 2-bromo-1-4-methoxyphenyl,p-methoxyphenacyl bromide,4'-methoxyphenacyl bromide,2-bromo-1-4-methoxyphenyl ethan-1-one,4-methoxyphenacyl bromide,bromomethyl 4-methoxyphenyl ketone,alpha-bromo-4-methoxyacetophenone,2-bromo-4-methoxyacetophenone
4965
2-bromo-1-(4-methoxyphenyl)ethanone
COC1=CC=C(C=C1)C(=O)CBr
Quantity 5g
Infrared Spectrum Authentic
Melting Point 69°C to 72°C
Packaging Glass bottle
Color White to Brown
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula CH3OC6H4COCH2Br
Beilstein 08, 89
Fieser 05,439
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most organic solvents
Formula Weight 229.07
Physical Form Crystals or Crystalline Powder
Percent Purity 98%
Chemical Name or Material 2-Bromo-4'-methoxyacetophenone
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 220-118-8