Learn More
Accela Chembio IncSupplier Diversity Partner 2-BROMO-4-METHOXYBENZALDE 5G  DFS Item
Manufacturer:  Accela Chembio IncSupplier Diversity Partner SY0263495G
2-BROMO-4-METHOXYBENZALDEHYDE, 5G, 43192-31-0, MFCD09788636, 97+%, RETEST PERIOD: 540DAYS, LIGHT SENSITIVE/AIR SENSITIVE/+4 c
Catalog No. 50-649-663