Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-Bromo-4-methoxyphenol 98.0+%, TCI America™
Click to view available options
Quantity:
5g
25g
Specifications
Melting Point | 43°C |
Boiling Point | 115°C |
Quantity | 5g |
Color | Undesignated |
Formula Weight | 203.04 |
Physical Form | Crystal-Powder at 20°C |
Percent Purity | ≥98.0% (T) |
Chemical Name or Material | 2-Bromo-4-methoxyphenol |
Safety and Handling
TSCA : No