Alfa Aesar™ 2'-Bromo-4'-methylacetanilide, 98%

Manufacturer: Alfa Aesar™ B2221306

Quantity 5g

 View more versions of this product

Catalog No. AAB2221306

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 614-83-5
    Molecular Formula C9H10BrNO
    Formula Weight 228.09
    MDL Number MFCD00017781
    Beilstein 2691019
    Chemical Name or Material 2'-Bromo-4'-methylacetanilide

    Specifications

    Formula Weight 228.09
    TSCA TSCA
    Molecular Formula C9H10BrNO
    MDL Number MFCD00017781
    CAS 614-83-5
    Melting Point 117° to 119°C
    Beilstein 2691019
    Quantity 5g
    Chemical Name or Material 2'-Bromo-4'-methylacetanilide
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit