Alfa Aesar™ 2-Bromo-4-methylpyridine, 96%

Manufacturer: Alfa Aesar™ B2525306

Quantity 5g

 View more versions of this product

Catalog No. AAB2525306

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 4926-28-7
    Molecular Formula C6H6BrN
    Formula Weight 172.03
    MDL Number MFCD00082590
    Beilstein 107331
    Synonym 2-Bromo-4-picoline
    Chemical Name or Material 2-Bromo-4-methylpyridine

    Specifications

    Formula Weight 172.03
    Molecular Formula C6H6BrN
    MDL Number MFCD00082590
    Synonym 2-Bromo-4-picoline
    Boiling Point 87°C (10mmHg)
    Quantity 5g
    Flash Point >110°C (230°F)
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H311-H332-H315-H319
    Toxic if swallowed.
    Toxic in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P261-P301+P310-P305+P351+P338-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 4926-28-7
    Beilstein 107331
    Chemical Name or Material 2-Bromo-4-methylpyridine
    Assay 96%

    GHS H Statement
    H301-H311-H332-H315-H319
    Toxic if swallowed.
    Toxic in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P261-P301+P310-P305+P351+P338-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit