Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-4-methylpyridine, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
10g
Packaging:
Glass bottle
4926-28-7
C6H6BrN
172.02
MFCD00082590
LSZMVESSGLHDJE-UHFFFAOYSA-N
2-bromo-4-picoline,2-bromo-4-methyl-pyridine,4-methyl-2-bromopyridine,pyridine, 2-bromo-4-methyl,2 bromo-4-methyl pyridine,2-bromo-4-methyl pyridine,pubchem1184,acmc-209kfi,2-bromo4-methylpyridine
2734087
2-bromo-4-methylpyridine
CC1=CC(=NC=C1)Br
4926-28-7
C6H6BrN
172.02
MFCD00082590
LSZMVESSGLHDJE-UHFFFAOYSA-N
2-bromo-4-picoline,2-bromo-4-methyl-pyridine,4-methyl-2-bromopyridine,pyridine, 2-bromo-4-methyl,2 bromo-4-methyl pyridine,2-bromo-4-methyl pyridine,pubchem1184,acmc-209kfi,2-bromo4-methylpyridine
2734087
2-bromo-4-methylpyridine
CC1=CC(=NC=C1)Br
Refractive Index 1.5600 to 1.563
Boiling Point 87°C (10.0mmHg)
Density 1.5450g/mL
Flash Point >109°C
Specific Gravity 1.545
Color Yellow
Quantity 1g
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Formula Weight 172.02
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material 2-Bromo-4-methylpyridine, 99%
Show More Show Less

GHS H Statement
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if inhaled.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor

GHS Signal Word: Warning

TSCA : TSCA