Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Bromo-4'-nitroacetophenone, 95%

Click to view available options
Quantity:
10g
50g
99-81-0
C8H6BrNO3
244.044
MFCD00007356
MBUPVGIGAMCMBT-UHFFFAOYSA-N
2-bromo-4'-nitroacetophenone,2-bromo-1-4-nitrophenyl ethanone,4-nitrophenacyl bromide,p-nitrophenacyl bromide,2-bromo-1-4-nitrophenyl ethan-1-one,alpha-bromo-p-nitroacetophenone,ethanone, 2-bromo-1-4-nitrophenyl,4'-nitrophenacyl bromide,alpha-bromo-4-nitroacetophenone,2-bromo-4-nitroacetophenone
66840
2-bromo-1-(4-nitrophenyl)ethanone
C1=CC(=CC=C1C(=O)CBr)[N+](=O)[O-]
99-81-0
C8H6BrNO3
244.044
MFCD00007356
MBUPVGIGAMCMBT-UHFFFAOYSA-N
2-bromo-4'-nitroacetophenone,2-bromo-1-4-nitrophenyl ethanone,4-nitrophenacyl bromide,p-nitrophenacyl bromide,2-bromo-1-4-nitrophenyl ethan-1-one,alpha-bromo-p-nitroacetophenone,ethanone, 2-bromo-1-4-nitrophenyl,4'-nitrophenacyl bromide,alpha-bromo-4-nitroacetophenone,2-bromo-4-nitroacetophenone
66840
2-bromo-1-(4-nitrophenyl)ethanone
C1=CC(=CC=C1C(=O)CBr)[N+](=O)[O-]
Quantity 50g
Melting Point 95°C to 98°C
UN Number UN1759
Beilstein 393567
Formula Weight 244.05
Percent Purity 95%
Chemical Name or Material 2-Bromo-4'-nitroacetophenone

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Hazard Class: 8; Packaging Group: II

EINECSNumber : 202-789-9

TSCA : Yes

Recommended Storage : Ambient temperatures