Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Bromo-4-nitrophenetole, 99%

Click to view available options
Quantity:
5g
25g
100g
58244-42-1
C8H8BrNO3
246.06
MFCD12547883
DAJMJBJIPXARBJ-UHFFFAOYSA-N
2-bromo-4-nitrophenetole,3-bromo-4-ethoxynitrobenzene,2-brom-4-nitrophenetol,acmc-209m3a,2-ethoxy-5-nitrobromobenzene
18787093
2-bromo-1-ethoxy-4-nitrobenzene
CCOC1=C(C=C(C=C1)[N+](=O)[O-])Br
58244-42-1
C8H8BrNO3
246.06
MFCD12547883
DAJMJBJIPXARBJ-UHFFFAOYSA-N
2-bromo-4-nitrophenetole,3-bromo-4-ethoxynitrobenzene,2-brom-4-nitrophenetol,acmc-209m3a,2-ethoxy-5-nitrobromobenzene
18787093
2-bromo-1-ethoxy-4-nitrobenzene
CCOC1=C(C=C(C=C1)[N+](=O)[O-])Br
Quantity 25g
Melting Point 94°C to 98°C
Formula Weight 246.06
Percent Purity 99%
Chemical Name or Material 2-Bromo-4-nitrophenetole

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger