Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Bromo-4'-phenylacetophenone, 97%

Click to view available options
Quantity:
5g
25g
100g
135-73-9
C14H11BrO
275.145
MFCD00000202
KGHGZRVXCKCJGX-UHFFFAOYSA-N
2-bromo-4'-phenylacetophenone,4-phenylphenacyl bromide,p-bromoacetylbiphenyl,p-phenylphenacyl bromide,bromomethyl p-biphenylyl ketone,ethanone, 1-1,1'-biphenyl-4-yl-2-bromo,2-bromo-1-4-phenylphenyl ethan-1-one,acetophenone, 2-bromo-4'-phenyl,alpha-bromo-p-phenylacetophenone,omega-bromo-4-phenylacetophenone
67282
2-bromo-1-(4-phenylphenyl)ethanone
C1=CC=C(C=C1)C2=CC=C(C=C2)C(=O)CBr
135-73-9
C14H11BrO
275.145
MFCD00000202
KGHGZRVXCKCJGX-UHFFFAOYSA-N
2-bromo-4'-phenylacetophenone,4-phenylphenacyl bromide,p-bromoacetylbiphenyl,p-phenylphenacyl bromide,bromomethyl p-biphenylyl ketone,ethanone, 1-1,1'-biphenyl-4-yl-2-bromo,2-bromo-1-4-phenylphenyl ethan-1-one,acetophenone, 2-bromo-4'-phenyl,alpha-bromo-p-phenylacetophenone,omega-bromo-4-phenylacetophenone
67282
2-bromo-1-(4-phenylphenyl)ethanone
C1=CC=C(C=C1)C2=CC=C(C=C2)C(=O)CBr
Quantity 5g
Melting Point 123°C to 127°C
UN Number UN3261
Beilstein 513838
Formula Weight 275.15
Percent Purity 97%
Chemical Name or Material 2-Bromo-4'-phenylacetophenone

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

TSCA : TSCA