Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-4-trifluoromethoxyaniline, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
175278-17-8
C7H5BrF3NO
256.02
MFCD00203478
ROSTYHNIIDIBEG-UHFFFAOYSA-N
2-bromo-4-trifluoromethoxy aniline,2-bromo-4-tirfluoromethoxyaniline,2-bromo-4-trifluoromethoxy-phenylamine,benzenamine, 2-bromo-4-trifluoromethoxy,2-bromo-4-trifluoromethoxy phenylamine,acmc-1c8k7,ksc495e4t,2-bromo-4 trifluoromethoxy aniline
688296
2-bromo-4-(trifluoromethoxy)aniline
C1=CC(=C(C=C1OC(F)(F)F)Br)N
175278-17-8
C7H5BrF3NO
256.02
MFCD00203478
ROSTYHNIIDIBEG-UHFFFAOYSA-N
2-bromo-4-trifluoromethoxy aniline,2-bromo-4-tirfluoromethoxyaniline,2-bromo-4-trifluoromethoxy-phenylamine,benzenamine, 2-bromo-4-trifluoromethoxy,2-bromo-4-trifluoromethoxy phenylamine,acmc-1c8k7,ksc495e4t,2-bromo-4 trifluoromethoxy aniline
688296
2-bromo-4-(trifluoromethoxy)aniline
C1=CC(=C(C=C1OC(F)(F)F)Br)N
Boiling Point 60°C to 64°C (0.3mmHg)
Density 1.6930g/mL
Flash Point 109°C
Specific Gravity 1.693
Refractive Index 1.5030 to 1.505
Packaging Glass bottle
Quantity 5g
Infrared Spectrum Authentic
Color Orange-Yellow to Yellow
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 256.02
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material 2-Bromo-4-trifluoromethoxyaniline
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

Recommended Storage : Product may darken on storage