Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Bromo-5-chloroanisole, 98+%

Click to view available options
Quantity:
5g
25g
174913-09-8
C7H6BrClO
221.478
MFCD03790889
CQGYLDZGJLVLMK-UHFFFAOYSA-N
2-bromo-5-chloroanisole,benzene, 1-bromo-4-chloro-2-methoxy,pubchem5265,acmc-1c20c,ksc497i0p,2-bromo-5-chloroanisole, 98+%,2-methoxy-4-chloro-1-bromobenzene,1-bromo-4-chloro-2-methoxy-benzene
17984845
1-bromo-4-chloro-2-methoxybenzene
COC1=C(C=CC(=C1)Cl)Br
174913-09-8
C7H6BrClO
221.478
MFCD03790889
CQGYLDZGJLVLMK-UHFFFAOYSA-N
2-bromo-5-chloroanisole,benzene, 1-bromo-4-chloro-2-methoxy,pubchem5265,acmc-1c20c,ksc497i0p,2-bromo-5-chloroanisole, 98+%,2-methoxy-4-chloro-1-bromobenzene,1-bromo-4-chloro-2-methoxy-benzene
17984845
1-bromo-4-chloro-2-methoxybenzene
COC1=C(C=CC(=C1)Cl)Br
Quantity 5g
Formula Weight 221.48
Percent Purity ≥98%
Chemical Name or Material 2-Bromo-5-chloroanisole