Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Bromo-5-chlorobenzaldehyde, 97%

Click to view available options
Quantity:
5g
25g
100g
174265-12-4
C7H4BrClO
219.462
MFCD00462870
IIISHLMCTDMUHH-UHFFFAOYSA-N
5-chloro-2-bromobenzaldehyde,benzaldehyde, 2-bromo-5-chloro,pubchem19849,acmc-1bwmm,ksc174i5n,2-bromo-5-chlorobenzaldehycle,iiishlmctdmuhh-uhfffaoysa
15391279
2-bromo-5-chlorobenzaldehyde
C1=CC(=C(C=C1Cl)C=O)Br
174265-12-4
C7H4BrClO
219.462
MFCD00462870
IIISHLMCTDMUHH-UHFFFAOYSA-N
5-chloro-2-bromobenzaldehyde,benzaldehyde, 2-bromo-5-chloro,pubchem19849,acmc-1bwmm,ksc174i5n,2-bromo-5-chlorobenzaldehycle,iiishlmctdmuhh-uhfffaoysa
15391279
2-bromo-5-chlorobenzaldehyde
C1=CC(=C(C=C1Cl)C=O)Br
Melting Point 64°C to 68°C (literature)
Color Yellow
Quantity 25g
Beilstein 10423891
Formula Weight 219.46
Physical Form Crystalline
Percent Purity 97%
Chemical Name or Material 2-Bromo-5-chlorobenzaldehyde

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures