Alfa Aesar™ 2-Bromo-5-chlorobenzoic acid, 98+%

Manufacturer: Alfa Aesar™ A1010306

Quantity 5g

 View more versions of this product

Catalog No. AAA1010306

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 21739-93-5
    Molecular Formula C7H4BrClO2
    Formula Weight 235.46
    MDL Number MFCD00013982
    UN Number UN2811
    Beilstein 2442261
    Chemical Name or Material 2-Bromo-5-chlorobenzoic acid

    Specifications

    Formula Weight 235.46
    Health Hazard 3 GHS P Statement
    P280h-P305+P351+P338-P309-P310
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Health Hazard 2 GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    MDL Number MFCD00013982
    CAS 21739-93-5
    Beilstein 2442261
    Molecular Formula C7H4BrClO2
    UN Number UN2811
    Melting Point 152° to 156°C
    Quantity 5g
    Odor Odorless
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    Chemical Name or Material 2-Bromo-5-chlorobenzoic acid
    Assay ≥98%

    GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280h-P305+P351+P338-P309-P310
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit