Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 2-Bromo-5-chloropyridine, 98%
Click to view available options
Quantity:
25g
100g
Description
Applications
It is an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields. It is also used in the synthesis of novel halopyridinylboronic acids and esters.
Solubility
Slightly soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
It is an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields. It is also used in the synthesis of novel halopyridinylboronic acids and esters.
Solubility
Slightly soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Quantity | 25g |
Boiling Point | 128°C (16mmHg) |
Melting Point | 65°C to 69°C (literature) |
Beilstein | 109098 |
Solubility Information | Slightly soluble in water. |
Formula Weight | 192.44 |
Physical Form | Solid |
Percent Purity | 98% |
Chemical Name or Material | 2-Bromo-5-chloropyridine |
Safety and Handling
GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H332-H315-H319-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: III
TSCA : No
Recommended Storage : Ambient temperatures