Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-5-chloropyridine, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
40473-01-6
C5H3BrClN
192.44
MFCD00234006
BZUUVQCSPHPUQA-UHFFFAOYSA-N
5-chloro-2-bromopyridine,2-bromo-5-chloro-pyridine,2-brom-5-chlorpyridin,pyridine, 2-bromo-5-chloro,pubchem1169,acmc-209jeb,2-bromo-5-chloro pyridine,ksc238i0d,tpc-py102,2-bromanyl-5-chloranyl-pyridine
817098
2-bromo-5-chloropyridine
C1=CC(=NC=C1Cl)Br
40473-01-6
C5H3BrClN
192.44
MFCD00234006
BZUUVQCSPHPUQA-UHFFFAOYSA-N
5-chloro-2-bromopyridine,2-bromo-5-chloro-pyridine,2-brom-5-chlorpyridin,pyridine, 2-bromo-5-chloro,pubchem1169,acmc-209jeb,2-bromo-5-chloro pyridine,ksc238i0d,tpc-py102,2-bromanyl-5-chloranyl-pyridine
817098
2-bromo-5-chloropyridine
C1=CC(=NC=C1Cl)Br
Color Beige to Brown-Yellow
Infrared Spectrum Authentic
Quantity 1g
Melting Point 65°C to 69°C
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 192.44
Physical Form Powder
Percent Purity ≥97.5%
Chemical Name or Material 2-Bromo-5-chloropyridine
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning