Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Bromo-5-cyanopyridine, 97%

Click to view available options
Quantity:
1g
5g
139585-70-9
C6H3BrN2
183.008
MFCD00234141
XHYGUDGTUJPSNX-UHFFFAOYSA-N
2-bromo-5-cyanopyridine,6-bromonicotinonitrile,2-bromo-5-pyridinecarbonitrile,5-cyano-2-bromopyridine,6-bromo-nicotinonitrile,6-bromo-3-pyridinecarbonitrile,3-pyridinecarbonitrile, 6-bromo,pubchem5963,2-bromo-5-cyano-pyridine,acmc-209cj9
5005718
6-bromopyridine-3-carbonitrile
C1=CC(=NC=C1C#N)Br
139585-70-9
C6H3BrN2
183.008
MFCD00234141
XHYGUDGTUJPSNX-UHFFFAOYSA-N
2-bromo-5-cyanopyridine,6-bromonicotinonitrile,2-bromo-5-pyridinecarbonitrile,5-cyano-2-bromopyridine,6-bromo-nicotinonitrile,6-bromo-3-pyridinecarbonitrile,3-pyridinecarbonitrile, 6-bromo,pubchem5963,2-bromo-5-cyano-pyridine,acmc-209cj9
5005718
6-bromopyridine-3-carbonitrile
C1=CC(=NC=C1C#N)Br
Quantity 5g
UN Number UN3439
Beilstein 8760368
Formula Weight 183.01
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 2-Bromo-5-cyanopyridine

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.